Learn More
BUCHI Drying Flask
SDP
Catalog No. 05405016 Shop All BUCHI Products
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-405-016 2000 mL
05-405-015 500 mL
05-405-004 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-016 Supplier BUCHI Supplier No. 011580
Add to Cart
Add to Cart
Pear shaped flasks with indents to reduce accumulations on the glass walls for increased efficiency in powder drying.
  • Indents reduce accumulation on the flask walls
  • Increased efficiency in powder drying
  • Standard Taper Joint: 24/40
Specifications
Material Borosilicate Glass (3.3)
Type Drying Flask
For Use With (Application) Powder Drying
For Use With (Equipment) Rotary Evaporators
Shape Pear
Standard Taper Joint Number Center 24/40
Volume (Metric) 2000 mL