Learn More
KPM ANALYTICS NORTH AMERICA CORPORATION KPM Analytics Drying Pad, Prevents Cross-Contamination, Supports Accurate Analytical Results, 1 EA
Catalog No. NC9316865
Encompass
The KPM Analytics Drying Pad ensures accurate analytical results by effectively drying components within SmartChem Series Automated Discrete Analyzers. It prevents cross-contamination between samples, thereby enhancing reliability in wet chemistry analysis. This critical consumable differentiates itself by playing an essential role in maintaining the integrity of measurements, making it an indispensable tool for any lab utilizing KPM Analytics technology.
Catalog No. NC9316865 Supplier KPM ANALYTICS NORTH AMERICA CORPORATION Supplier No. 901003800
The KPM Analytics Drying Pad ensures accurate analytical results by effectively drying components within SmartChem Series Automated Discrete Analyzers. It prevents cross-contamination between samples, thereby enhancing reliability in wet chemistry analysis. This critical consumable differentiates itself by playing an essential role in maintaining the integrity of measurements, making it an indispensable tool for any lab utilizing KPM Analytics technology.