Learn More
METTLER TOLEDO™ Drying Pump DryPro
Catalog No. 13940055 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Reversible:
No
Catalog No. Reversible
13-940-055 No
1 options
This item is not returnable. View return policy
Catalog No. 13-940-055 Supplier METTLER TOLEDO™ Supplier No. 30474899
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Ensure Fast and Efficient Drying
Connect the DryPro to thoroughly dry the measuring cell of Excellence Density Meters, even in humid environments, before an adjustment or measurement
TRUSTED_SUSTAINABILITY
Specifications
Reversible No
Includes Silica gel