Learn More
Cole-Parmer Modular stainless steel drying rack, 50 white pegs
Supplier:  Cole-Parmer 6703510
Encompass_Preferred
Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds. Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable. Integral catch drain directs all drainage into the drip troughprevents accidental drainage to the back of the rack. Racks come with 6"L pegs, SS drain tube, and wall mounting bracket.
Catalog No. NC0604141