Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Drying Rack Replacement Pegs
GREENER_CHOICE

Catalog No. 03448349
Change view
Click to view available options
Color:
Blue, Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-448-349 Blue, Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-448-349 Supplier Heathrow Scientific™ Supplier No. 120793
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Five long and five short replacement pegs for the Lab Drying Rack Set

  • SureGrip Hook with Patent-pending design to securely grip glassware into place

Specifications

Autoclavable Not Autoclavable
Color Blue, Green
Holds Lab Glassware
Material ABS
Positions 1
Shape Flat
Stackable No
Sterility Non-sterile
For Use With (Equipment) Lab Drying Rack
Quantity 5 Long Pegs and 5 Short Pegs
Dimensions (L x W x H) Short pegs: 5.4 x 0.4 x 0.78 in.; Long pegs: 6.9 x 0.4 x 0.78 in.
Product Type Replacement Peg
Shelves None
Height (Metric) 5 cm
Array 1 x 1
Width (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.