Learn More
Cole-Parmer Modular stainless steel drying rack, 60 white pegs
Catalog No. NC1497566 Shop All Cole-Parmer Products
Encompass_Preferred
Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds. Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable. Integral catch drain directs all drainage into the drip trough—prevents accidental drainage to the back of the rack.
Catalog No. NC1497566 Supplier Cole-Parmer Supplier No. 6703520
Removable "snap-in" pegs and add-on accessories let you turn these drying racks into customized work stations in just seconds. Corrosion-resistant 304 stainless steel (SS) racks and polypropylene pegs are easy to clean. Autoclavable. Integral catch drain directs all drainage into the drip trough—prevents accidental drainage to the back of the rack.