Learn More
Ace Glass, Inc. DRYING TOWER DRYING TOWER
SDP
Catalog No. 50979476
Encompass_Preferred
Drying Tower with a 45/50 inner joint and a #11 thread for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-979-476 Supplier Ace Glass, Inc. Supplier No. 781830
Drying Tower with a 45/50 inner joint and a #11 thread for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.