Learn More
Chemglass Life Sciences Drying Tower, 250mm Overall Height
SDP
Catalog No. NC1420894
Encompass_Preferred
Cylindrical shaped drying tower has a 34/28 ground glass stopper. The stopper has a hole coinciding with a serrated hose connection sealed in the outer portion of the stopper neck and a second hose connection near the base of the cylinder. A hex base is sealed to the bottom of the tower for added stability.
Catalog No. NC1420894 Supplier Chemglass Life Sciences Supplier No. CG129205
Cylindrical shaped drying tower has a 34/28 ground glass stopper. The stopper has a hole coinciding with a serrated hose connection sealed in the outer portion of the stopper neck and a second hose connection near the base of the cylinder. A hex base is sealed to the bottom of the tower for added stability.