Learn More
Chemglass Life Sciences Drying Tube, 75° Degree, 24/40 Inner Joint, # 3 Rubber Stopper
SDP
Catalog No. NC1420931
Encompass_Preferred
The 75 angle drying tube is for use in fume hoods where space is at a minimum. Supplied complete with rubber stopper, glass inlet tube, and a lower inner joint.
Catalog No. NC1420931 Supplier Chemglass Life Sciences Supplier No. CG129501
The 75 angle drying tube is for use in fume hoods where space is at a minimum. Supplied complete with rubber stopper, glass inlet tube, and a lower inner joint.