Learn More
Medchemexpress LLC Dryocrassin ABBA | 12777-70-7 | 820.83 | 5 MG
SDP
Catalog No. 5000388526
Encompass_Preferred
Dryocrassin ABBA (Dryocrassin) is an orally active phloroglucinol derivative extracted from *Phyllopteris officinalis*. It demonstrates antitumor and antiviral activity, and induces apoptosis of human hepatocellular carcinoma cells via a mitochondrial pathway mediated by Caspase.
  • Possesses antitumor activity
  • Exhibits antiviral activity
  • Induces apoptosis in human hepatocellular carcinoma cells via a mitochondrial pathway
  • Inhibits S. aureus SrtA (sortase A) activity
Catalog No. 50-003-88526 Supplier Medchemexpress LLC Supplier No. HYN05305MG
Add to Cart
Add to Cart
Dryocrassin ABBA (Dryocrassin) is an orally active phloroglucinol derivative extracted from *Phyllopteris officinalis*. It demonstrates antitumor and antiviral activity, and induces apoptosis of human hepatocellular carcinoma cells via a mitochondrial pathway mediated by Caspase.
  • Possesses antitumor activity
  • Exhibits antiviral activity
  • Induces apoptosis in human hepatocellular carcinoma cells via a mitochondrial pathway
  • Inhibits S. aureus SrtA (sortase A) activity