Learn More
Slice™ Drywall Knife
Catalog No. 17010325
Change view
Click to view available options
Blade Material:
Zirconium Oxide
Catalog No. Blade Material
17-010-325 Zirconium Oxide
1 options
This item is not returnable. View return policy
Catalog No. 17-010-325 Supplier Slice™ Supplier No. 10582
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Drywall Knife was designed with advice from drywall professionals. Its blade change screw head fits your tape measure hook. The installed finger-friendly™ blade with Never-Lost™ safety cover keeps you working and safe.
The Slice Drywall Knife was designed with advice from drywall professionals, explicitly created for scoring drywall. The handle narrows at the blade end, allowing for easier handling as you hold a tape measure to maintain a consistent edge distance while scoring. However, it's versatile enough to use as an all-purpose utility knife.
From scoring drywall to slicing heavy materials and plastic sheeting, Slice makes people and places safer. Learn more below.
  • Durable aluminum handle with safety cover
  • Easy to clean
  • 100% zirconium oxide blade that never rusts
  • Blade lasts on average 11.2 times longer than metal blades
  • Easy-to-use ambidextrous design
Great for:
  • Scoring drywall
  • Cutting plastic sheeting and cardboard
  • Slicing heavy-duty packaging
  • Cutting screens with a pointed-tip blade
Specifications
Blade Material Zirconium Oxide
Product Type Knife
For Use With (Application) Scoring drywall; Cutting plastic sheeting and cardboard; Slicing heavy-duty packaging; Cutting screens with a pointed-tip blade
Length (English) Blade 1.34 in.
Length (Metric) Blade 34 mm
Size 6.1 x 1.26 x 0.91 in.
Sterility Non-sterile
Includes Slice Blade 10524
Handle Material Alloy Aluminum
Color Silver, Orange