Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ DS-02 Matrix Standard Kit, for 3500/3730/SeqStudio™/SeqStudio™ Flex

Catalog No. 4323014
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 Runs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-230-14 8 Runs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-230-14 Supplier Applied Biosystems™ Supplier No. 4323014
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Contains dye-labeled oligonucleotides used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit

The dye-labeled oligonucleotides included in the Matrix Standard DS-02 set (dR110, dR6G, dTAMRA™, dROX™, and LIZ™) are used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit.

The dye-labeled oligonucleotides included in the Matrix Standard DS-02 set (dR110, dR6G, dTAMRA, dROX, and LIZ) are used to generate the 'multicomponent matrix' required for the SNaPshot Multiplex Kit. The Data Collection software utilizes the multicomponent matrix to automatically correct for the spectral overlap in samples labeled with DS-02 dyes. The kit consists of one tube of matrix standard which is sufficient for a minimum of eight array runs (on a 24 capillary) or two array runs (on a 96 capillary).

Matrix standards do not need to be run with every set of sample injections. The standard only needs to be run once in order to generate a matrix file which is then applied to samples run under similar conditions. For more information on the use of matrix standards, refer to the instrument User Manual or Getting Started Guide.

Used with SNaPshot Multiplex Kit on 3500/3500xl, 3730/3730xl, SeqStudio, and SeqStudio Flex series genetic analyzers.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains dye-labeled oligonucleotides used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit. Store at 2 - 8°C. Do not freeze.
Format Kit
For Use With (Equipment) SeqStudio™ Genetic Analyzer, 3730 Series Genetic Analyzers, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Label or Dye LIZ, dRhodamine 6G, dRhodamine ROX, dRhodamine TAMRA
Product Type DS-02 Matrix Standard Kit
Quantity 8 Runs
Shipping Condition Wet Ice
What is a spectral calibration?

A spectral calibration is an algorithm applied to raw data, which converts it into the component 4 or 5 dye data stored in the sample files. A spectral is created for a specific dye set (combination of dyes), array type (4 or 16 capillaries), and array length (36cm or 50cm). It is used to correct for the natural overlap of the fluorescent dyes.

Which fragment analysis matrix standards can I use for my Applied Biosystems 3130 Series instrument?

DS-02 (Dye Set E5), DS-30 (Dye Set D), DS-31 (Dye Set D), DS-32 (Dye Set F), and DS-33 (Dye Set G5) are all supported on the Applied Biosystems 3130 Series systems. Please refer to the Applied Biosystems 3130/3130xl Genetic Analyzers Getting Started Guide (http://tools.thermofisher.com/content/sfs/manuals/cms_041468.pdf) for more information.

I am using only one dye for fragment analysis but I see peaks in other colors below my peak of interest. Why is this?

If the peaks in other colors are directly below the peak of interest, the issue could be that the fluorescent dye being used is not part of the selected dye set, the spectral calibration needs to be performed, or the peaks are offscale. Confirm that the dye set selected on the instrument is compatible with the dye being used, run a new spectral calibration if the correct dye set has been selected and, if the signal intensity is too high, decrease sample concentration during PCR or when preparing samples for electrophoresis.

How many SNPs can be multiplexed with the SNaPshot Multiplex Kit?

The SNaPshot Multiplex Kit is designed to interrogate up to ten single nucleotide polymorphisms (SNPs) at known locations on one to ten DNA templates in a single tube.

How do I design my primers for the SNaPshot Multiplex Kit?

Follow these recommendations for designing and evaluating primers:

- Primers included in a single reaction need to differ significantly in lengths in order to avoid overlap between the final SNaPshot products. A difference of 4-6 nucleotides between primer lengths is recommended as a starting point (5-7 nucleotides if running on POP-7 Polymer).
- The length of a primer can be modified by the addition of non-homologous polynucleotides at the 5′ end. Since the recommended annealing temperature for a SNaPshot control primer is 50 degrees C, the melting temperature for the complementary region between any primer and its corresponding template should be at least 50 degrees C.
- Poly (dT), poly (dA), poly (dC), and poly (dGACT) are 5′ non-homologous tails which are predicted to have minimal secondary structures. They have all been used successfully. Generally the signal patterns are not affected by the kinds of tails that are used. The 5′ poly (dT) tails however may interfere with the addition of 3′ ddA.
- The mobility of an oligonucleotide in capillary electrophoresis is determined by its size, nucleotide composition, and dye. Thus the effect of nucleotide composition on mobility can be significant when the primer is short. We strongly recommend that you test primers shorter than 36 nucleotides before being multiplexed to ensure that the final products are spatially resolved when analyzed on the instrument.
- Check primers for possible extendable hairpin structures within each primer and for extendable dimer formation between primers.
- HPLC purification of primers is recommended for oligonucleotides longer than 30 nucleotides. Heterogeneous primer mixtures containing mixed molecular weight oligonucleotides may yield undesired products that will confuse analysis.

For additional suggestions please refer to Appendix A of the SNaPshot Multiplex Kit manual (https://tools.thermofisher.com/content/sfs/manuals/cms_041203.pdf).

What dye set does the SNaPshot Multiplex Kit use?

The SNaPshot Multiplex Kit uses the E5 dye set which contains the dR110, dR6G, dTAMRA, and dROX dyes, and uses LIZ dye-labeled size standard.

Can I switch dyes during a fragment analysis project?

We do not recommend that you switch dyes in the middle of a project. Switching dyes may result in a shift in the allele sizes as dyes differ in their mobility. If dyes are switched in the middle of a project, it will be necessary to identify the size shift for all the markers/targets in the amplicon product range. The size shift will be consistent throughout the project and it is only necessary to make the adjustment once.

What is a dye set?

A dye set is a combination of dyes that have minimum spectral overlap, which will allow you to multiplex products of similar size using different dyes. Although the size ranges may overlap, the use of a different dye will allow the software to easily distinguish between these products.

Am I able to use dyes other than the recommended dyes for primer labeling prior to PCR amplification for fragment analysis?

We recommend using only Applied Biosystems dyes, as we provide spectral calibration reagents that have been optimized for our dye sets. Non-Applied Biosystems dyes have variable emission spectra and also require a spectral calibration generated for the specific dyes in use to correct for the spectral overlap between the dyes. You are responsible for obtaining the appropriate spectral calibration reagents and for optimizing custom dye sets to ensure that the dye labels do not affect PCR efficiency. The use of dyes outside of the recommended dye sets can result in pull-up and pull-down peaks, which may make allele-calling challenging.

I would like to use another company's kit to generate fragments for my fragment analysis run. Is this possible?

It is possible to use another company's kit. However, the vendor should be contacted to determine if they have protocol(s) for their kit(s) on the specific capillary electrophoresis (CE) instrument (specifically, the model you intend to use), and if they have reagents to run a spectral calibration or matrix run on the CE instrument with compatibility to the dyes that are part of the kit(s) in question.

Do you have any fragment analysis-based kits for AFLP analysis?

Unfortunately, we do not have fragment analysis-based kits for AFLP analysis. General information regarding AFLP can be found here (https://www.thermofisher.com/us/en/home/life-science/sequencing/fragment-analysis/amplified-fragment-length-polymorphism-aflp-analysis.html).

What kits are available for SNP genotyping by fragment analysis?

We offer the SNaPshot Multiplex Kit for SNP genotyping. Please refer to our overview of SNP Genotyping by Fragment Analysis (https://www.thermofisher.com/us/en/home/life-science/sequencing/dna-sequencing/snp-genotyping-variant-detection-sequencing/snp-genotyping-fragment-analysis.html) for additional information.

Which fragment analysis applications can I run on the capillary electrophoresis (CE) instruments?

There are several fragment analysis applications that can be run on the CE instruments, such as: microsatellite analysis, SNP genotyping, fingerprinting, and relative fluorescence quantitation. Please see the DNA Fragment Analysis by Capillary Electrophoresis Guide for more details about these applications (http://tools.thermofisher.com/content/sfs/manuals/4474504.pdf).

What reagents do I need to start a fragment analysis project?

In order to determine the reagents needed, it will be necessary to gather initial information such as the following:
- What is the minimum and maximum amplicon size range?
- Will there be one target per sample or multiple targets (singleplex versus multiplex on the capillary electrophoresis instrument)?
- Is there overlap among the size of amplicons?

This initial information will help identify the spectral calibration needed for the capillary electrophoresis instrument, the fluorescent dye used to label the primer, and the size standard. Additional information can be found in the “Experimental Design” section of the DNA Fragment Analysis by Capillary Electrophoresis Guide (http://tools.thermofisher.com/content/sfs/manuals/4474504.pdf).

When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.