Learn More
Medchemexpress LLC DS-1001b | 1898207-64-1 | >99.7% | 10 MG
SDP
Catalog No. 5000340726
Encompass_Preferred
DS-1001b is an orally active, blood-brain permeable, and potent inhibitor of mutant IDH-1 (isocitrate dehydrogenase-1). It exhibits antitumor activity.
  • Orally active
  • Blood-brain permeable
  • Potent inhibitor of mutant IDH-1
  • Exhibits antitumor activity
Catalog No. 50-003-40726 Supplier Medchemexpress LLC Supplier No. HY12954510MG
DS-1001b is an orally active, blood-brain permeable, and potent inhibitor of mutant IDH-1 (isocitrate dehydrogenase-1). It exhibits antitumor activity.
  • Orally active
  • Blood-brain permeable
  • Potent inhibitor of mutant IDH-1
  • Exhibits antitumor activity