Learn More
Medchemexpress LLC DS-1001b | 1898207-64-1 | 99.7% | 100 MG
SDP
Catalog No. 5000338759
Encompass_Preferred
DS-1001b is an orally active, blood-brain permeable, potent isocitrate dehydrogenase-1 (IDH-1) mutant inhibitor. This compound exhibits antitumor activity.
  • Orally active
  • Blood-brain permeable
  • Potent IDH-1 mutant inhibitor
  • Exhibits antitumor activity
Catalog No. 50-003-38759 Supplier Medchemexpress LLC Supplier No. HY129545100MG
DS-1001b is an orally active, blood-brain permeable, potent isocitrate dehydrogenase-1 (IDH-1) mutant inhibitor. This compound exhibits antitumor activity.
  • Orally active
  • Blood-brain permeable
  • Potent IDH-1 mutant inhibitor
  • Exhibits antitumor activity