Learn More
Medchemexpress LLC DS-1001b | 1898207-64-1 | 99.7% | 50 MG
SDP
Catalog No. 5000338496
Encompass_Preferred
DS-1001b is an orally active, blood-brain permeable, potent IDH-1 (isocitrate dehydrogenase-1) mutant inhibitor with antitumor activity. It targets IDH-1 and shows promise in various biological activities.
  • Orally active and blood-brain permeable
  • Potent IDH-1 mutant inhibitor
  • Exhibits antitumor activity
  • Inhibits proliferation of IDH1-mutated chondrosarcoma cells
  • Upregulates SOX9, RUNX2, COL2A1, COL10A1, and ACAN expression
  • Induces G1 phase arrest
  • Damages subcutaneous tumor growth in xenografted mice
Catalog No. 50-003-38496 Supplier Medchemexpress LLC Supplier No. HY12954550MG
DS-1001b is an orally active, blood-brain permeable, potent IDH-1 (isocitrate dehydrogenase-1) mutant inhibitor with antitumor activity. It targets IDH-1 and shows promise in various biological activities.
  • Orally active and blood-brain permeable
  • Potent IDH-1 mutant inhibitor
  • Exhibits antitumor activity
  • Inhibits proliferation of IDH1-mutated chondrosarcoma cells
  • Upregulates SOX9, RUNX2, COL2A1, COL10A1, and ACAN expression
  • Induces G1 phase arrest
  • Damages subcutaneous tumor growth in xenografted mice