Learn More
Medchemexpress LLC HY-129545 5mg Medchemexpress, DS-1001b CAS:1898207-64-1 Purity:>98%
SDP
Catalog No. 501969809
Encompass_Preferred
Medchemexpress, HY-129545 5mg DS-1001b CAS:1898207-64-1 DS-1001b is a mutant IDH-1 (Isocitrate Dehydrogenase-1) inhibitor extracted from patent WO2016052697A1, Example 168, and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9809 Supplier Medchemexpress LLC Supplier No. HY1295455MG
Medchemexpress, HY-129545 5mg DS-1001b CAS:1898207-64-1 DS-1001b is a mutant IDH-1 (Isocitrate Dehydrogenase-1) inhibitor extracted from patent WO2016052697A1, Example 168, and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.