Learn More
Medchemexpress LLC DS-1971a | 1450595-86-4 | 99.3% | 50 MG
SDP
Catalog No. 5000340344
Encompass_Preferred
DS-1971a is a potent, selective, and orally active NaV1.7 inhibitor, with IC50s of 22.8 nM for hNaV1.7 and 59.4 nM for mNaV1.7. It exerts analgesic effects and addresses thermal hyperalgesia and mechanical hypersensitivity.
  • Potent, selective, and orally active NaV1.7 inhibitor
  • Inhibitory concentration (IC50) of 22.8 nM for hNaV1.7
  • Exerts analgesic effects
  • Soluble in DMSO at 100 mg/mL
  • Powder stable for 3 years at -20°C and 2 years at 4°C
  • Stable in solvent for 2 years at -80°C and 1 year at -20°C
  • Targets sodium channel membrane transporter/ion channel
  • Exhibits a favorable toxicological profile
Catalog No. 50-003-40344 Supplier Medchemexpress LLC Supplier No. HY13118250MG
DS-1971a is a potent, selective, and orally active NaV1.7 inhibitor, with IC50s of 22.8 nM for hNaV1.7 and 59.4 nM for mNaV1.7. It exerts analgesic effects and addresses thermal hyperalgesia and mechanical hypersensitivity.
  • Potent, selective, and orally active NaV1.7 inhibitor
  • Inhibitory concentration (IC50) of 22.8 nM for hNaV1.7
  • Exerts analgesic effects
  • Soluble in DMSO at 100 mg/mL
  • Powder stable for 3 years at -20°C and 2 years at 4°C
  • Stable in solvent for 2 years at -80°C and 1 year at -20°C
  • Targets sodium channel membrane transporter/ion channel
  • Exhibits a favorable toxicological profile