Learn More
Medchemexpress LLC HY-131182 5mg Medchemexpress, DS-1971a CAS:1450595-86-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1311825MG
Encompass_Preferred
Medchemexpress, HY-131182 5mg DS-1971a CAS:1450595-86-4 DS-1971a is a potent, selective, and orally active NaV1.7 inhibitor, with IC50s of 22.8 and 59.4 nM for hNaV1.7 and mNaV1.7, respectively. DS-1971a exerts analgesic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1268