Learn More
Cole-Parmer Traceable Panel-Mount Thermometer with Calibration, -50 to 300°C (-58 to 572°F); 1 Remote Probe
Catalog No. NC1677820 Shop All Cole-Parmer Products
Encompass_Preferred
Panel-mount Remote Probe Thermometer features a 10-ft flexible cord for measuring temperatures from a distance. Versatile panel-mount module snaps into place or can be used as a stand-alone unit. Model has an large easy-view LCD display, and records minimum and maximum readings over time. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC1677820 Supplier Cole-Parmer Supplier No. 9020526
Panel-mount Remote Probe Thermometer features a 10-ft flexible cord for measuring temperatures from a distance. Versatile panel-mount module snaps into place or can be used as a stand-alone unit. Model has an large easy-view LCD display, and records minimum and maximum readings over time. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.