Learn More
Medchemexpress LLC Ds08210767 | 2376334-75-5 | 99.4% | 513.67 | C31H39N5O2 | 10MG
SDP
Catalog No. 5000239187
Encompass_Preferred
DS08210767 is a small-molecule antagonist of the parathyroid hormone type 1 receptor (PTHR1) with a reported in vitro IC50 of 90 nM. It is described as orally bioavailable and is supplied for research use only; not for therapeutic or human use. Manufacturer-provided analytical documentation is available.
Catalog No. 50-002-39187 Supplier Medchemexpress LLC Supplier No. HY12587910MG
DS08210767 is a small-molecule antagonist of the parathyroid hormone type 1 receptor (PTHR1) with a reported in vitro IC50 of 90 nM. It is described as orally bioavailable and is supplied for research use only; not for therapeutic or human use. Manufacturer-provided analytical documentation is available.