Learn More
Medchemexpress LLC DS08210767 | 2376334-75-5 | 99.4% | 513.67 | C31H39N5O2 | 25MG
SDP
Catalog No. 5000235528
Encompass_Preferred
DS08210767 is a small-molecule antagonist of the parathyroid hormone receptor 1 (PTHR1) for research use. It is reported to be orally bioavailable and shows an in vitro IC50 of 90 nM against PTHR1. The compound is supplied as a light yellow to yellow solid with high reported purity and is intended for biochemical and cellular studies; follow manufacturer storage recommendations.
Catalog No. 50-002-35528 Supplier Medchemexpress LLC Supplier No. HY12587925MG
DS08210767 is a small-molecule antagonist of the parathyroid hormone receptor 1 (PTHR1) for research use. It is reported to be orally bioavailable and shows an in vitro IC50 of 90 nM against PTHR1. The compound is supplied as a light yellow to yellow solid with high reported purity and is intended for biochemical and cellular studies; follow manufacturer storage recommendations.