Learn More
Medchemexpress LLC DS08210767 | 2376334-75-5 | 99.4% | 513.67 | C31H39N5O2 | 5MG
SDP
Catalog No. 5000235021
Encompass_Preferred
DS08210767 is a small-molecule, orally bioavailable antagonist of the parathyroid hormone receptor 1 (PTHR1) with a reported IC50 of 90 nM. It is supplied for research use only and is provided as a light yellow solid intended for laboratory applications.
  • Orally bioavailable PTHR1 antagonist with an IC50 of 90 nM.
  • High chemical purity of 99.4%.
  • Molecular weight 513.67 and chemical formula C31H39N5O2.
  • Supplied as a light yellow solid suitable for laboratory research.
  • Provided in a 5 mg quantity for experimental use.
Catalog No. 50-002-35021 Supplier Medchemexpress LLC Supplier No. HY1258795MG
DS08210767 is a small-molecule, orally bioavailable antagonist of the parathyroid hormone receptor 1 (PTHR1) with a reported IC50 of 90 nM. It is supplied for research use only and is provided as a light yellow solid intended for laboratory applications.
  • Orally bioavailable PTHR1 antagonist with an IC50 of 90 nM.
  • High chemical purity of 99.4%.
  • Molecular weight 513.67 and chemical formula C31H39N5O2.
  • Supplied as a light yellow solid suitable for laboratory research.
  • Provided in a 5 mg quantity for experimental use.