Learn More
Medchemexpress LLC DS08210767 | 2376334-75-5 | 99.4% | 513.67 g/mol | C31H39N5O2 | 100MG
SDP
Catalog No. 5000236478
Encompass_Preferred
DS08210767 is a small-molecule antagonist of parathyroid hormone receptor 1 (PTHR1), described as highly potent and orally bioavailable with an IC50 of 90 nM. It is provided as a research-grade, well-characterized compound suitable for receptor pharmacology and oral in vivo studies.
  • High potency (IC50 of 90 nM) for PTHR1 inhibition.
  • Orally bioavailable, enabling oral dosing in pharmacology studies.
  • High purity (99.4%) as verified by certificate of analysis.
  • Molecular weight 513.67 g/mol; formula C31H39N5O2.
  • Analytical data available: COA, HNMR, and LCMS.
  • Supplied in multiple milligram pack sizes for experimental flexibility.
Catalog No. 50-002-36478 Supplier Medchemexpress LLC Supplier No. HY125879100MG
DS08210767 is a small-molecule antagonist of parathyroid hormone receptor 1 (PTHR1), described as highly potent and orally bioavailable with an IC50 of 90 nM. It is provided as a research-grade, well-characterized compound suitable for receptor pharmacology and oral in vivo studies.
  • High potency (IC50 of 90 nM) for PTHR1 inhibition.
  • Orally bioavailable, enabling oral dosing in pharmacology studies.
  • High purity (99.4%) as verified by certificate of analysis.
  • Molecular weight 513.67 g/mol; formula C31H39N5O2.
  • Analytical data available: COA, HNMR, and LCMS.
  • Supplied in multiple milligram pack sizes for experimental flexibility.