Learn More
TARGETMOL CHEMICALS INC DS08210767 5MG
SDP
Catalog No. 502581573
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DS08210767 is a highly potent orally bioavailable PTHR1 antagonist with IC50 of 90 nM. purity: 98%
Catalog No. 50-258-1573 Supplier TARGETMOL CHEMICALS INC Supplier No. T111015MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DS08210767 is a highly potent orally bioavailable PTHR1 antagonist with IC50 of 90 nM. purity: 98%