Learn More
Medchemexpress LLC HY-130251 5mg Medchemexpress, DS18561882 CAS:2227149-22-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302515MG
Encompass_Preferred
Medchemexpress, HY-130251 5mg DS18561882 CAS:2227149-22-4 DS18561882 is a highly potent, isozyme-selective methylenetetrahydrofolate dehydrogenase 2 (MTHFD2) inhibitor with an IC50 value of 0.0063 μM. DS18561882 also has inhibitory effect on MTHFD1 (IC50=0.57 μM). DS18561882 exhibits a good oral pharmacokinetic profile. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8946