Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynex™ DS2™ System

Catalog No. 23333001
Change view
Click to view available options
Analytical Method:
Photometric
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analytical Method
23-333-001 Photometric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Appropriate precautions must be taken when working with biohazardous material.
Catalog No. 23-333-001 Supplier Dynex™ Supplier No. 62010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Appropriate precautions must be taken when working with biohazardous material.

Automated ELISA processor with 2 microplate incubators and barcode reader.

*
  • Featuring automation, accuracy, immediacy and responsiveness - the must-haves of demanding near-patient-care settings and lower-throughput labs
  • Reliable, cost-effective and easy to use/ maintain package
  • Completely automated open system
  • Well fill verification
  • 2 independent plate incubators
  • Unique dispense synchronization
  • 96 on-board samples
  • Up to 12 different assays simultaneously
  • Automatic data reduction
  • Easy-to-Use DS-Matrix Software
  • Disposable sample and reagent tips
  • Specifications

    Type Open System
    Description 2 plates system
    For Use With (Application) ELISA Assays
    Sample Dilution (96) 12 x 8 deep-well strips Maximum dilution: 1 to 5,000
    Throughput Assay dependent
    Analytical Method Photometric
    Capacity 100 samples on-board
    Certifications/Compliance EN 61326-1:2013EN 61326-2-6:2013 IEC/EN 61010-1:2010 Ed.3 IEC/EN 61010-2-010:2014 Ed.3 IEC/EN 61010-2-101 issued 2015/08/14 Ed. 2 CSA C22.2 No. 61010-1:2012 Ed.3 CSA C22.2 No. 61010-2-101:2015 Ed.2 RoHS Compliant
    Includes One year warranty against defects in parts, materials, and workmanship
    Wavelength 405/690 nm
    Depth (English) 27 in.
    Depth (Metric) 68 cm
    Height (English) 26 in.
    Height (Metric) 66 cm
    Width (English) 21 in.
    Width (Metric) 54 cm
    Electrical Requirements 100/240 V auto-switching
    Frequency 50/60 Hz
    Data Management Features DS-Matrix™ Software
    Operating System Windows 7/ Windows 10
    Measurement Method Absorbance
    Weight (English) 105 lb.
    Weight (Metric) 48 kg
    Show More Show Less

    DYNEX makes no representations, warranties or performance claims with respect to the performance of Agility for any specific application, including clinical application or for the use of the Agility system with any reagents, assays or other products.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.