Learn More
Medchemexpress LLC DS28120313 5mg | 2146177-09-3 | 5MG
SDP
Catalog No. 5000242872
Encompass_Preferred
DS28120313 (compound 32) is an orally hepcidin production inhibitor with an IC50 of 0 093 M[1
Catalog No. 50-002-42872 Supplier Medchemexpress LLC Supplier No. HY1079805MG
Add to Cart
Add to Cart
DS28120313 (compound 32) is an orally hepcidin production inhibitor with an IC50 of 0 093 M[1