Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ DS360 High Capacity Hand Sanitizing Wipes Station
SDP

Catalog No. 19044090 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-044-090 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-044-090 Supplier PURELL™ Supplier No. 911401SLVHSW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Premium, high-capacity floor stand dispenser for PURELL™ wipes.

  • Durable steel frame with optimized lid design to prevent rusting
  • Integrated waste receptacle keeps the surrounding area clean
  • 360 degree design makes wipes accessible from any angle
  • The REDIPUL™ Nozzle's durable construction lasts over 3 years, helping to reduce roping from nozzle wear
  • The REDIPUL™ Nozzle virtually eliminates wipe fallback and helps provide a readily accessible wipe every time
  • Throw away fewer dried-out wipes with the REDIPUL™ Nozzle's patent-pending re-wicking technology
  • Storage compartment allows you to store additional wipe refills for easy maintenance
  • Weighted base keeps stand in place
  • Customizable panels can be used as billboards for your message and branding
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sanitizing Cloth Dispenser
Color Black
Length (Metric) 15.5 cm
Width (Metric) 15.5 cm
Height (Metric) 39.6 cm
Holds Hand Sanitizer
Material Plastic
Mounting Freestanding
No. of Compartments 1
Product Style Top Loading
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.