Learn More
Medchemexpress LLC Ds45500853 | 2735803-28-6 | 99.2% | 284.35 g·mol⁻1 | C18H20O3 | 5 MG
SDP
Catalog No. 5000262226
Encompass_Preferred
DS45500853 is a small-molecule agonist of estrogen-related receptor alpha (ERRα) used in preclinical research to investigate metabolic pathways and disorders such as type 2 diabetes. It inhibits the interaction between RIP140 corepressor peptide and the ERRα ligand-binding domain with reported activity in the submicromolar to micromolar range and is supplied as a high-purity research compound.
  • Agonist of estrogen-related receptor alpha (ERRα).
  • Inhibits RIP140-ERRα corepressor binding with reported activity (IC50 ~0.80 μM).
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO for typical assay preparations.
  • Available in small milligram quantities for preclinical studies.
Catalog No. 50-002-62226 Supplier Medchemexpress LLC Supplier No. HY1322055MG
DS45500853 is a small-molecule agonist of estrogen-related receptor alpha (ERRα) used in preclinical research to investigate metabolic pathways and disorders such as type 2 diabetes. It inhibits the interaction between RIP140 corepressor peptide and the ERRα ligand-binding domain with reported activity in the submicromolar to micromolar range and is supplied as a high-purity research compound.
  • Agonist of estrogen-related receptor alpha (ERRα).
  • Inhibits RIP140-ERRα corepressor binding with reported activity (IC50 ~0.80 μM).
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO for typical assay preparations.
  • Available in small milligram quantities for preclinical studies.