Learn More
Medchemexpress LLC DS86760016 | 1853176-89-2 | 99.3% | 243.45 | C9H11BClNO4 | 50MG
SDP
Catalog No. 5000236355
Encompass_Preferred
DS86760016 is a leucyl-tRNA synthetase (LeuRS) inhibitor used for research on antibacterial mechanisms; it shows low-micromolar enzyme inhibition and in vitro activity against multidrug-resistant Gram-negative bacteria, and is supplied as a high-purity solid with recommended sealed, moisture-free storage for research use.
  • Potent LeuRS inhibition with reported IC50 values of ~0.16-0.62 μM.
  • Antibacterial activity against Gram-negative pathogens with MICs of 0.25-2 μg/mL.
  • High reported purity for reliable experimental use.
  • Solid form; off-white to light brown appearance and sealed, dry storage recommended.
  • Available in small research package sizes suitable for laboratory studies.
Catalog No. 50-002-36355 Supplier Medchemexpress LLC Supplier No. HY12467950MG
DS86760016 is a leucyl-tRNA synthetase (LeuRS) inhibitor used for research on antibacterial mechanisms; it shows low-micromolar enzyme inhibition and in vitro activity against multidrug-resistant Gram-negative bacteria, and is supplied as a high-purity solid with recommended sealed, moisture-free storage for research use.
  • Potent LeuRS inhibition with reported IC50 values of ~0.16-0.62 μM.
  • Antibacterial activity against Gram-negative pathogens with MICs of 0.25-2 μg/mL.
  • High reported purity for reliable experimental use.
  • Solid form; off-white to light brown appearance and sealed, dry storage recommended.
  • Available in small research package sizes suitable for laboratory studies.