Learn More
Cayman Chemical D-Saccharic Acid 1,4-lactone (hydrate), Purity: ≥95%, CAS: 61278-30-6, 250 mg
SDP
Catalog No. 502984704
Encompass_Preferred
An inhibitor of β-glucuronidase (IC50 = 45 µM for the human enzyme)
Catalog No. 50-298-4704 Supplier Cayman Chemical Supplier No. 18896250 MG
An inhibitor of β-glucuronidase (IC50 = 45 µM for the human enzyme)