Learn More
Enzo Life Sciences DsbA (E. coli), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ALX201153C025
Encompass_Preferred
Alternative name: Thiol disulfide interchange protein DsbA. Purity: ≥99%. Formulation: Liquid. In water. Source: Produced in E. coli. Long Term Storage: -80°C. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles. Applications: WB .
Catalog No. 50-200-7941