Learn More
Enzo Life Sciences DsbC (E. coli), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX201268C100
Encompass_Preferred
Alternative name: Thiol disulfide interchange protein DsbC. Concentration: 1mg/ml. Purity: ≥95% (SDS-PAGE). MW: ~23.5kDa. Formulation: Liquid. In 20mM TRIS, pH 7.5, containing 2mM EDTA. Source: Produced in E. coli. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7962