Learn More
Fauske & Associates, a Division of Stone & Webster, LLC DSC-F11-RAM accessory for Differential Scanning Calorimetry, 10 mm
Catalog No. NC3761441
Encompass
The FAUSKE AND ASSOCIATES INC DSC-F11-RAM is a specialized accessory designed for use with Differential Scanning Calorimetry (DSC) systems. This device is instrumental in sample preparation and handling, enabling thermal hazard assessment and material property evaluation. As part of a suite of solutions for process safety, this accessory exemplifies high-quality manufacturing standards and extensive industry expertise.
  • Designed for use with DSC systems
  • Facilitates sample preparation and thermal hazard assessments
  • Utilizes insights from process safety expertise
  • Compatible with F11 Toggle Press
  • Has a dimension of 10 mm
Catalog No. NC3761441 Supplier Fauske & Associates, a Division of Stone & Webster, LLC Supplier No. DSCF11RAM
The FAUSKE AND ASSOCIATES INC DSC-F11-RAM is a specialized accessory designed for use with Differential Scanning Calorimetry (DSC) systems. This device is instrumental in sample preparation and handling, enabling thermal hazard assessment and material property evaluation. As part of a suite of solutions for process safety, this accessory exemplifies high-quality manufacturing standards and extensive industry expertise.
  • Designed for use with DSC systems
  • Facilitates sample preparation and thermal hazard assessments
  • Utilizes insights from process safety expertise
  • Compatible with F11 Toggle Press
  • Has a dimension of 10 mm