Learn More
Anton Paar DSC Autosampler
Catalog No. 5000580546
Encompass_Preferred
DSC autosampler for automatic change of sample and reference crucibles. Comprises 70 sample positions and 8 reference positions.Reference positions can be used for reference crucibles as well as calibration samples.Crucibles can be dropped post-analysis automatically into a crucible waste container.Includes: Autosampler Calibration Tool (266628)
Catalog No. 50-005-80546 Supplier Anton Paar Supplier No. 261476
Please contact your sales representative for a quote or further ordering instructions.
DSC autosampler for automatic change of sample and reference crucibles. Comprises 70 sample positions and 8 reference positions.Reference positions can be used for reference crucibles as well as calibration samples.Crucibles can be dropped post-analysis automatically into a crucible waste container.Includes: Autosampler Calibration Tool (266628)