Learn More
TA Instruments DSC DTA Certified Temperature Calibration Kit, Indium and Adamantane Reference Materials, Traceable Temperature and Enthalpy Calibration, Documentation and Accessories Included
Catalog No. 5000580679 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments DSC DTA Certified Temperature Calibration Kit contains indium and adamantane certified reference materials with documentation and accessories for traceable temperature and enthalpy calibration of DSC and DTA cells.
Catalog No. 50-005-80679 Supplier TA Instruments Supplier No. 915060.901
Waters TA Instruments DSC DTA Certified Temperature Calibration Kit contains indium and adamantane certified reference materials with documentation and accessories for traceable temperature and enthalpy calibration of DSC and DTA cells.