Learn More
TA Instruments DSC Hermetic Die Set White, Die Set Replacement for Tzero Sample Press for Hermetic Sealing Standard Aluminum and Gold Sample Pans, Consists of Three Dies
Catalog No. 5000580732 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments DSC Hermetic Die Set is a replacement white die set for the Tzero Sample Press. It is designed for hermetic sealing DSC Standard aluminum and gold sample pans. The set includes three dies, a lower die, and two upper dies for encapsulating moisture sensitive materials effectively.
Catalog No. 50-005-80732 Supplier TA Instruments Supplier No. 901608.902
Waters TA Instruments DSC Hermetic Die Set is a replacement white die set for the Tzero Sample Press. It is designed for hermetic sealing DSC Standard aluminum and gold sample pans. The set includes three dies, a lower die, and two upper dies for encapsulating moisture sensitive materials effectively.