Learn More
TA Instruments DSC High Volume Die Set Yellow, Die Set Optional for Tzero Sample Press for Sealing High Volume Stainless Steel Pans Lids and Seals, Consists of Two Dies
Catalog No. 5000580907 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments DSC High Volume Die Set is an optional yellow die set for the Tzero Sample Press. It is designed for sealing samples using high volume stainless steel pans, lids, and seals. The set consists of two dies, a lower and upper die, ensuring secure encapsulation of pressure sensitive materials.
Catalog No. 50-005-80907 Supplier TA Instruments Supplier No. 901608.905
Waters TA Instruments DSC High Volume Die Set is an optional yellow die set for the Tzero Sample Press. It is designed for sealing samples using high volume stainless steel pans, lids, and seals. The set consists of two dies, a lower and upper die, ensuring secure encapsulation of pressure sensitive materials.