Learn More
Anton Paar DSC Piercing Device
Catalog No. 5000580544
Encompass_Preferred
Lid piercing accessory for Julia DSC autosampler. Allows piercinghermetically sealed crucibles prior to measurement. The cruciblesmust be closed with a special type of lid (Mat. No. 266337).
Catalog No. 50-005-80544 Supplier Anton Paar Supplier No. 261129
Lid piercing accessory for Julia DSC autosampler. Allows piercinghermetically sealed crucibles prior to measurement. The cruciblesmust be closed with a special type of lid (Mat. No. 266337).