Learn More
TA Instruments DSC Standard Die Set Green, Die Set Replacement for Tzero Sample Press for Crimp Sealing DSC Standard Aluminum and Gold Sample Pans and Lids, Consists of Three Dies
Catalog No. 5000580840 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments DSC Standard Die Set is a replacement green die set for the Tzero Sample Press. It is designed for crimp sealing DSC Standard aluminum and gold sample pans and lids. The set includes three dies, a lower die, and flat and concave upper dies for encapsulating various sample forms with precision and consistency.
Catalog No. 50-005-80840 Supplier TA Instruments Supplier No. 901608.901
Waters TA Instruments DSC Standard Die Set is a replacement green die set for the Tzero Sample Press. It is designed for crimp sealing DSC Standard aluminum and gold sample pans and lids. The set includes three dies, a lower die, and flat and concave upper dies for encapsulating various sample forms with precision and consistency.