Learn More
TA Instruments DSC Tzero Die Set Black, Die Set Replacement for Tzero Sample Press for Crimp Sealing Tzero Aluminum Pans and Lids, Consists of Three Dies
Catalog No. 5000580592 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments DSC Tzero Die Set is a replacement black die set for the Tzero Sample Press. It is designed for crimp sealing Tzero aluminum pans and lids. The set consists of three dies, including a lower die and flat and concave upper dies, to encapsulate various sample forms with accuracy.
Catalog No. 50-005-80592 Supplier TA Instruments Supplier No. 901608.903
Waters TA Instruments DSC Tzero Die Set is a replacement black die set for the Tzero Sample Press. It is designed for crimp sealing Tzero aluminum pans and lids. The set consists of three dies, including a lower die and flat and concave upper dies, to encapsulate various sample forms with accuracy.