Learn More
Aviva Systems Biology Corp DSC3 Antibody - N-terminal region (ARP45195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DSC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DSC3. Alternate names: CDHF3, DSC, DSC1, DSC2, DSC4, HT-CPApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58980