Learn More
Braintree Scientific ""Blue Wax"" Dissection Tray, 9"" x 6.5"" x 1"", 5/pk
SDP
Catalog No. NC9031079
Encompass
"Blue Wax" Dissection TrayAdd a dissection tray with a wax base pinning surface for detailed dissections! Specially formulated to provide proper consistency to allow pins to stick firmly and be easily removed. Dissection tray uses a completely environmentally safe wax product instead of the "black wax" like product used in most other dissection trays. MSDS sheet for this product lists no toxic chemicals and no fire, health or reactivity hazards with this product. "Blue Wax" can be heated to renew surface and dissection tray can be reused several times. **Sold for research purposes only and not for human use**
Catalog No. NC9031079 Supplier Braintree Scientific Supplier No. DTW5
"Blue Wax" Dissection TrayAdd a dissection tray with a wax base pinning surface for detailed dissections! Specially formulated to provide proper consistency to allow pins to stick firmly and be easily removed. Dissection tray uses a completely environmentally safe wax product instead of the "black wax" like product used in most other dissection trays. MSDS sheet for this product lists no toxic chemicals and no fire, health or reactivity hazards with this product. "Blue Wax" can be heated to renew surface and dissection tray can be reused several times. **Sold for research purposes only and not for human use**