Learn More
Agilent Technologies dsDNA 905 Reagent Kit, Qualitative, 35 to 500 bp, 1,000 Samples
Catalog No. NC1964479
Encompass
The dsDNA 905 Reagent Kit provides automated and accurate sizing and relative quantification of DNA fragments. Designed for use with Fragment Analyzer systems, this kit is suitable for applications such as the analysis of PCR amplicons, simple sequence repeats (SSRs), microsatellites, and genotyping. The kit contains specially formulated gels, markers, and ladders to ensure reliable performance across a broad range of fragment sizes.
  • Provides a sizing range of 35 bp to 500 bp with ± 5 % accuracy.
  • Supports a concentration range of 0.5 ng/μL to 50 ng/μL.
  • Requires a low sample volume of 2 μL.
  • Delivers high resolution with 2 % CV precision.
  • Enables short separation times for efficient analysis.
  • For research use only; not for use in diagnostic procedures.
Catalog No. NC1964479 Supplier Agilent Technologies Supplier No. DNF905K1000
The dsDNA 905 Reagent Kit provides automated and accurate sizing and relative quantification of DNA fragments. Designed for use with Fragment Analyzer systems, this kit is suitable for applications such as the analysis of PCR amplicons, simple sequence repeats (SSRs), microsatellites, and genotyping. The kit contains specially formulated gels, markers, and ladders to ensure reliable performance across a broad range of fragment sizes.
  • Provides a sizing range of 35 bp to 500 bp with ± 5 % accuracy.
  • Supports a concentration range of 0.5 ng/μL to 50 ng/μL.
  • Requires a low sample volume of 2 μL.
  • Delivers high resolution with 2 % CV precision.
  • Enables short separation times for efficient analysis.
  • For research use only; not for use in diagnostic procedures.