Learn More
Zymo Research Corporation dsDNA Shearase™ Plus 50 U (1 U/ul) + DCC-5 (D4013) '**'
SDP
Catalog No. 502037196
Encompass_Preferred
dsDNA Shearase™ Plus is an endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5-phosphate and 3-hydroxyl termini. It has a particularly strong preference for double-stranded DNA (dsDNA) and generates random-ended DNA fragments of the desired size in a single step. Sequencing data demonstrates that dsDNA Shearase™ Plus does not introduce any detectable bias in the sequencing library preparation. This enzyme is compatible with low volume inputs thus minimizing sample loss. Digested DNA is easily purified in 10 l with recommended DNA Clean & Concentrator™ technology making it ideal for use in end modification (linker & adapter) procedures and other applications.
Catalog No. 50-203-7196 Supplier Zymo Research Corporation Supplier No. E201950
dsDNA Shearase™ Plus is an endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5-phosphate and 3-hydroxyl termini. It has a particularly strong preference for double-stranded DNA (dsDNA) and generates random-ended DNA fragments of the desired size in a single step. Sequencing data demonstrates that dsDNA Shearase™ Plus does not introduce any detectable bias in the sequencing library preparation. This enzyme is compatible with low volume inputs thus minimizing sample loss. Digested DNA is easily purified in 10 l with recommended DNA Clean & Concentrator™ technology making it ideal for use in end modification (linker & adapter) procedures and other applications.