Learn More
Zymo Research Corporation dsDNA Shearase™ Plus 50 U (1 U/ul)
SDP
Catalog No. 50444523
Encompass_Preferred
dsDNA Shearase Plus is an endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5’-phosphate and 3’-hydroxyl termini. It has a particularly strong preference for dsDNA and generates random-ended DNA fragments of the desired size in a single step. This enzyme is compatible with low volume inputs, thus minimizing sample loss.
Catalog No. 50-444-523 Supplier Zymo Research Corporation Supplier No. E201850
dsDNA Shearase Plus is an endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5’-phosphate and 3’-hydroxyl termini. It has a particularly strong preference for dsDNA and generates random-ended DNA fragments of the desired size in a single step. This enzyme is compatible with low volume inputs, thus minimizing sample loss.