Learn More
Biotium AccuClear dsDNA Standard, 25 ng/uL
Catalog No. 501964548
Encompass_Preferred
The AccuClear dsDNA Standard is a 25 ng/uL dsDNA standard, also provided in the AccuClear Ultra High Sensitivity dsDNA Quantitation Kits. It contains dsDNA from calf thymus, 25 ng/uL in 10 mM Tris pH 7.5, 1 mM EDTA, 2 mM sodium azide. Potential applications include making custom DNA standard curves for quantitation.
Catalog No. 50-196-4548 Supplier Biotium Supplier No. 31029C
The AccuClear dsDNA Standard is a 25 ng/uL dsDNA standard, also provided in the AccuClear Ultra High Sensitivity dsDNA Quantitation Kits. It contains dsDNA from calf thymus, 25 ng/uL in 10 mM Tris pH 7.5, 1 mM EDTA, 2 mM sodium azide. Potential applications include making custom DNA standard curves for quantitation.