Learn More
Cayman Chemical DSedoheptulose7phosphate 10mg
SDP
Catalog No. 502985303
Encompass_Preferred
An intermediate in the pentose phosphate pathway, carbon fixation in photosynthetic organisms, and the biosynthesis of lipopolysaccharide, amino acids, secondary metabolites, and antibiotics
Catalog No. 50-298-5303 Supplier Cayman Chemical Supplier No. 2134410 MG
An intermediate in the pentose phosphate pathway, carbon fixation in photosynthetic organisms, and the biosynthesis of lipopolysaccharide, amino acids, secondary metabolites, and antibiotics