Learn More
CPC Scientific Glp-His-Trp-Ser-Tyr-DSer(tBu)-Leu-Arg-Pro-(Aza)Gly-NH2 25MG
SDP
Supplier:  CPC Scientific LHRH003C
Encompass
Goserelin, a synthetic peptide analog of LHRH, is used in the treatment of prostate cancer, advanced breast cancer and in the management of endometriosis.SEQUENCE: Glp-His-Trp-Ser-Tyr-DSer(tBu)-Leu-Arg-Pro-(Aza)Gly-NH2ONE-LETTER SEQUENCE: Glp-HWSY-DS(tBu)-LRP-(Aza)G-NH2MOLECULAR FORMULA: C60H86N18O14MOLECULAR WEIGHT: 1269.43STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [145781-92-6]SYNONYMS: Zoladex?RESEARCH AREA: HormonalREFERENCES:A.Wells et al., Clin. Cancer Res., 8, 1251 (2002)A.Rody et al., Expert Rev. Anticancer Ther., 5, 591 (2005)H.Lepor, Rev. Urol., 7 (Suppl. 5), S3 (2005)Merck Index, 14th ed., No. 4526, (2006)M.Roach 3rd and A.Izaguirre, Expert Opin. Pharmacother., 8, 257 (2007)R.M.Herrmann et al., Rad. Oncol., 2, 31 (2007)
SKU(s): LHRH-003A, LHRH-003B, LHRH-003C
Catalog No. 50-210-9649