Learn More
Cayman Chemical Desethoxy Quetiapine (hydrochloride), Purity: ≥95%, CAS: 329218-14-6, 10 mg
SDP
Catalog No. 502909557
Encompass_Preferred
An active metabolite of quetiapine; binds to dopamine D2 receptors (IC50 = 1,330 nM); reduces apomorphine-induced climbing in mice; reduces apomorphine-induced swimming deficits in mice
Catalog No. 50-290-9557 Supplier Cayman Chemical Supplier No. 3541010 MG
An active metabolite of quetiapine; binds to dopamine D2 receptors (IC50 = 1,330 nM); reduces apomorphine-induced climbing in mice; reduces apomorphine-induced swimming deficits in mice